ENA321764413 Enamine

5-methyl-2-(pyrrolidin-1-yl)benzamide

  • MDL number MFCD18838782
  • Cas Number : 1306604-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76373

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76373
Availability details for
Quantity
  • Availability
  • Add to Cart