ENA321764414 Enamine

3,4-diaminobutanoic acid dihydrochloride

  • MDL number MFCD09836079
  • Cas Number : 109754-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76422

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76422
Availability details for
Quantity
  • Availability
  • Add to Cart