ENA321764416 Enamine

ethyl 2-(dimethylamino)-4-methyl-1,3-thiazole-5-carboxylate

  • MDL number MFCD18838785
  • Cas Number : 58350-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76443

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76443
Availability details for
Quantity
  • Availability
  • Add to Cart