ENA321764418 Enamine

2-(chloromethyl)-5-(propan-2-yl)-1,3-oxazole

  • MDL number MFCD18838787
  • Cas Number : 731851-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76458
Availability details for
Quantity
  • Availability
  • Add to Cart