ENA321764424 Enamine

3-(3-chlorophenyl)-5-ethenyl-1,2,4-oxadiazole

  • MDL number MFCD18917257
  • Cas Number : 1311318-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76486
Availability details for
Quantity
  • Availability
  • Add to Cart