ENA321764428 Enamine

1-[3-(chlorosulfonyl)-4-methylphenyl]-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD18838796
  • Cas Number : 1306605-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114...116
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76510

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76510
Availability details for
Quantity
  • Availability
  • Add to Cart