ENA321764431 Enamine

(5-cyclopropyl-1,2,4-oxadiazol-3-yl)(phenyl)methanamine hydrochloride

  • MDL number MFCD18838799
  • Cas Number : 1306603-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76523

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76523
Availability details for
Quantity
  • Availability
  • Add to Cart