ENA321764432 Enamine

4-(aminomethyl)-N-(1-phenylethyl)oxan-4-amine

  • MDL number MFCD18838801
  • Cas Number : 1306605-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76536

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76536
Availability details for
Quantity
  • Availability
  • Add to Cart