ENA321764433 Enamine

2,2,2-trifluoroethyl 2-[(morpholin-4-yl)methyl]pyrrolidine-1-carboxylate

  • MDL number MFCD18838802
  • Cas Number : 1306605-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76538
Availability details for
Quantity
  • Availability
  • Add to Cart