ENA321764441 Enamine

5-(2-bromophenyl)-1,2,4-oxadiazole

  • MDL number MFCD18838811
  • Cas Number : 1306604-33-0
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76614
Availability details for
Quantity
  • Availability
  • Add to Cart