ENA321764442 Enamine

ethyl 5H,6H,7H,8H-imidazo[1,2-a]pyrimidine-2-carboxylate

  • MDL number MFCD18838812
  • Cas Number : 143696-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171...173
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76615

Safety Information

msds https://www.enaminestore.com/catalog/EN300-76615
Availability details for
Quantity
  • Availability
  • Add to Cart