ENA321764444 Enamine

2-[3-(aminomethyl)phenoxy]acetonitrile hydrochloride

  • MDL number MFCD18838814
  • Cas Number : 1306603-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76652
Availability details for
Quantity
  • Availability
  • Add to Cart