ENA321764445 Enamine

tert-butyl 4-[(hydrazinecarbonyl)amino]piperidine-1-carboxylate

  • MDL number MFCD18917260
  • Cas Number : 1311317-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76658
Availability details for
Quantity
  • Availability
  • Add to Cart