ENA321764454 Enamine

octahydroindolizin-2-amine

  • MDL number MFCD18838826
  • Cas Number : 80220-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76687

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76687
Availability details for
Quantity
  • Availability
  • Add to Cart