ENA321764457 Enamine

2-amino-2-(2-bromophenyl)propanoic acid hydrochloride

  • MDL number MFCD18838828
  • Cas Number : 1306605-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-76700

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76700
Availability details for
Quantity
  • Availability
  • Add to Cart