ENA321764458 Enamine

2-(2-chloro-4-fluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD18838829
  • Cas Number : 874285-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76702

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76702
Availability details for
Quantity
  • Availability
  • Add to Cart