ENA321764475 Enamine

4-(4-ethylpiperazin-1-yl)-3-fluorobenzoic acid hydrochloride

  • MDL number MFCD18838849
  • Cas Number : 1306604-53-4
  • Molecular Weight : 288.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76734
Availability details for
Quantity
  • Availability
  • Add to Cart