ENA321764477 Enamine

(2S)-2-amino-3-methyl-1-(pyrrolidin-1-yl)butan-1-one hydrochloride

  • MDL number MFCD18838850
  • Cas Number : 115201-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-179
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76747
Availability details for
Quantity
  • Availability
  • Add to Cart