ENA321764492 Enamine

2-(diethylamino)-2-[3-(trifluoromethyl)phenyl]acetic acid hydrochloride

  • MDL number MFCD18838865
  • Cas Number : 1306602-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-76776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76776
Availability details for
Quantity
  • Availability
  • Add to Cart