ENA321764509 Enamine

2-(1H-pyrazol-4-yl)piperidine

  • MDL number MFCD18838882
  • Cas Number : 1306606-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76812
Availability details for
Quantity
  • Availability
  • Add to Cart