ENA321764513 Enamine

2-(3-tert-butylphenoxy)ethan-1-amine hydrochloride

  • MDL number MFCD18838886
  • Cas Number : 1306605-41-3
  • Molecular Weight : 229.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-76822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76822
Availability details for
Quantity
  • Availability
  • Add to Cart