ENA321764532 Enamine

[3-(pyridin-4-yl)-1,2,4-oxadiazol-5-yl]methanol hydrochloride

  • MDL number MFCD18838906
  • Cas Number : 1306603-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76898
Availability details for
Quantity
  • Availability
  • Add to Cart