ENA321764541 Enamine

4-{[6-(trifluoromethyl)pyridin-2-yl]oxy}aniline hydrochloride

  • MDL number MFCD18838914
  • Cas Number : 1306605-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-76971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76971
Availability details for
Quantity
  • Availability
  • Add to Cart