ENA321764550 Enamine

N-[4-(chloromethyl)-1,3-thiazol-2-yl]-N-(2-methoxyethyl)acetamide

  • MDL number MFCD18838923
  • Cas Number : 1306605-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-76995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-76995
Availability details for
Quantity
  • Availability
  • Add to Cart