ENA321764553 Enamine

1-[2-(propan-2-yl)-1,3-thiazol-4-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD18838926
  • Cas Number : 1306603-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77002

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77002
Availability details for
Quantity
  • Availability
  • Add to Cart