ENA321764555 Enamine

2-[(5-acetyl-2-methoxyphenyl)methanesulfinyl]acetic acid

  • MDL number MFCD18838928
  • Cas Number : 1306603-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77005

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77005
Availability details for
Quantity
  • Availability
  • Add to Cart