ENA321764557 Enamine

ethyl 1-amino-2,3-dihydro-1H-indene-1-carboxylate hydrochloride

  • MDL number MFCD18838930
  • Cas Number : 1306606-03-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77015

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77015
Availability details for
Quantity
  • Availability
  • Add to Cart