ENA321764562 Enamine

1-(5-amino-2,3-dihydro-1H-isoindol-2-yl)ethan-1-one

  • MDL number MFCD18838935
  • Cas Number : 722444-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77024

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-77024
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart