ENA321764581 Enamine

7-oxabicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD18838957
  • Cas Number : 19800-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76-77
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77106

Safety Information

signalword Warning
pcodes P264,P270,P301+P312,P330,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-77106
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart