ENA321764583 Enamine

ethyl 5-(methylamino)-1,3-thiazole-4-carboxylate

  • MDL number MFCD18917266
  • Cas Number : 1311316-82-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77120
Availability details for
Quantity
  • Availability
  • Add to Cart