ENA321764589 Enamine

2,2,2-trifluoroethyl N-[2-(piperidin-1-yl)ethyl]carbamate hydrochloride

  • MDL number MFCD18838962
  • Cas Number : 1306605-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77147

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77147
Availability details for
Quantity
  • Availability
  • Add to Cart