ENA321764596 Enamine

3-chloro-4-(pyridin-4-yloxy)benzoic acid hydrochloride

  • MDL number MFCD18838968
  • Cas Number : 1306604-54-5
  • Molecular Weight : 286.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-77207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77207
Availability details for
Quantity
  • Availability
  • Add to Cart