ENA321764598 Enamine

1-ethyl-1,2,3,4-tetrahydroisoquinoline hydrochloride

  • MDL number MFCD18838970
  • Cas Number : 103861-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77215
Availability details for
Quantity
  • Availability
  • Add to Cart