ENA321764603 Enamine

1-(azetidin-3-yl)-1H-imidazole dihydrochloride

  • MDL number MFCD18838975
  • Cas Number : 153836-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77248

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77248
Availability details for
Quantity
  • Availability
  • Add to Cart