ENA321764619 Enamine

6-(aminomethyl)-5,7-dimethyl-1H,2H-pyrazolo[1,5-a]pyrimidin-2-one dihydrochloride

  • MDL number MFCD18838989
  • Cas Number : 1306605-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77333

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77333
Availability details for
Quantity
  • Availability
  • Add to Cart