ENA321764621 Enamine

2-amino-1-(furan-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD18917270
  • Cas Number : 88352-86-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77341
Availability details for
Quantity
  • Availability
  • Add to Cart