ENA321764624 Enamine

(2-fluorophenyl)(5-methyl-1,2,4-oxadiazol-3-yl)methanamine hydrochloride

  • MDL number MFCD18838991
  • Cas Number : 1306604-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77354
Availability details for
Quantity
  • Availability
  • Add to Cart