ENA321764625 Enamine

5-chloro-2-methoxy-N-methylaniline hydrochloride

  • MDL number MFCD18838992
  • Cas Number : 1306606-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156...158
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77355
Availability details for
Quantity
  • Availability
  • Add to Cart