ENA321764626 Enamine

3-ethyl-4,5-dihydro-1,2-oxazole-5-carbonitrile

  • MDL number MFCD18838993
  • Cas Number : 66376-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77358

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77358
Availability details for
Quantity
  • Availability
  • Add to Cart