ENA321764633 Enamine

2-[4-(2-aminoethoxy)phenyl]acetonitrile hydrochloride

  • MDL number MFCD18838999
  • Cas Number : 1306603-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77411
Availability details for
Quantity
  • Availability
  • Add to Cart