ENA321764636 Enamine

3-amino-1-phenylpiperidin-2-one

  • MDL number MFCD18839002
  • Cas Number : 1233344-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77417

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77417
Availability details for
Quantity
  • Availability
  • Add to Cart