ENA321764646 Enamine

2-amino-5-methylhexanoic acid hydrochloride

  • MDL number MFCD18839013
  • Cas Number : 57224-44-9
  • Molecular Weight : 181.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-77459

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77459
Availability details for
Quantity
  • Availability
  • Add to Cart