ENA321764649 Enamine

3-(pyrrolidin-1-yl)oxan-4-amine dihydrochloride

  • MDL number MFCD18917277
  • Cas Number : 1311313-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77477

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77477
Availability details for
Quantity
  • Availability
  • Add to Cart