ENA321764651 Enamine

1-[2-(2-methylphenyl)-1,3-thiazole-4-sulfonyl]piperazine

  • MDL number MFCD18917278
  • Cas Number : 1311315-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77491

Safety Information

msds https://www.enaminestore.com/catalog/EN300-77491
Availability details for
Quantity
  • Availability
  • Add to Cart