ENA321764652 Enamine

2-chloro-4-(1H-imidazol-1-yl)pyridine

  • MDL number MFCD14702705
  • Cas Number : 1209458-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77496
Availability details for
Quantity
  • Availability
  • Add to Cart