ENA321764653 Enamine

3-amino-3-phenylprop-2-enethioamide

  • Cas Number : 1057326-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77498
Availability details for
Quantity
  • Availability
  • Add to Cart