ENA321764654 Enamine

3-(5-bromothiophen-2-yl)-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD18839016
  • Cas Number : 1306605-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77500
Availability details for
Quantity
  • Availability
  • Add to Cart