ENA321764655 Enamine

5-ethyl-3-methyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD18839017
  • Cas Number : 1306604-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-77501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77501
Availability details for
Quantity
  • Availability
  • Add to Cart