ENA321764657 Enamine

1-benzylpiperazine-2-carbonitrile

  • MDL number MFCD18839019
  • Cas Number : 947726-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77530
Availability details for
Quantity
  • Availability
  • Add to Cart