ENA321764659 Enamine

2-(piperazin-1-yl)propanamide dihydrochloride

  • MDL number MFCD18839021
  • Cas Number : 1306602-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-77533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-77533
Availability details for
Quantity
  • Availability
  • Add to Cart